Would you like to log in again?
HyFlex® CR+ Cut-Resistant Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyFlex®
205656 205655 205679 205659 205658 205657
89041-826CS 4279.88 USD InStock
89041-826 80090-006 80090-008 80090-000 80090-002 80090-004
HyFlex® CR+ Cut-Resistant Gloves, Ansell
Gloves
SType 11-501 gloves feature cut protection for applications such as automotive assembly or handling sheet metal, glass, or small sharp parts.
  • Great cut protection
  • Elasticized wrist
  • Slightly coarse finish
Gloves are constructed of nitrile-dipped foam and a unique Kevlar® fabric for superior cut resistance and dexterity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty