Would you like to log in again?
Whizard® Defender Armguard, Wells Lamont
Supplier: Wells Lamont
Whizard®
133683 133680
80090-657EA 65.42 USD InStock
80090-657 80090-653
Whizard® Defender Armguard, Wells Lamont
Workwear
The Whizard Defender armguards are made of high performance yarns and stainless steel.
  • High performance yarn and stainless steel construction
  • Clip provided
  • ANSI level 4
The 22" blue armguards feature a clip.
They are ANSI Level 4 armguards.
Certifications: ANSI Cut Level 4.
Ordering information: This armguard is sold in multiples of six pieces.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty