Would you like to log in again?
MetalGuard® Heavy Weight 1880 Cut Resistant Gloves, Wells Lamont
Supplier: Wells Lamont
Metalguard®
1880M 1880S 1880L 1880XL
80091-418EA 26.67 USD InStock
80091-418 80091-416 80091-414 80091-412
MetalGuard® Heavy Weight 1880 Cut Resistant Gloves, Wells Lamont
Gloves
MetalGuard® heavy weight gloves form to hands for comfortability.
  • Highest level of cut protection within the MetalGuard® line
  • Continuous knit conforms to the hand
MetalGuard® heavy weight gloves ensuring a close, secure fit. Additional reinforcement in the thumb crotch area increases wear life. Gloves are prewashed and preshrunk for a comfortable fit.
Certifications: ANSI Cut Level 5, A6.
Ordering information: These gloves are sold by the dozen pair.
Caution: WARNING: Cut resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty