Would you like to log in again?
Jomac® Terry Cloth Welder Glove, Loop Out, Duck Cuff, Wells Lamont
Supplier: Wells Lamont
628FR
80091-474EA 20.32 USD InStock
80091-474
Jomac® Terry Cloth Welder Glove, Loop Out, Duck Cuff, Wells Lamont
Gloves
Jomac terry cloth welder glove resists heat up to 250 °F.
  • Duck gauntlet cuff
  • Clute cut
  • Blue
  • Washing machine friendly
Users can clean the blue terry cloth glove in a washing machine.
Ordering information: Gloves are sold by the dozen pairs.
Caution: The degree of heat resistance a glove may offer is directly related to the duration of exposure and weight of the object being handled.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty