Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Jomac® Extra Heavy Weight Terry Cloth Gloves, Loop Out, Wells Lamont

Supplier: Wells Lamont
636HRLFR
80091-480EA 43.43 USD InStock
80091-480
Jomac® Extra Heavy Weight Terry Cloth Gloves, Loop Out, Wells Lamont
Gloves
These terry cloth gloves resist heat up to 450 °F.

  • Gauntlet cuff, 5"
  • Ambidextrous
  • Loop-out terry cloth
  • Cotton lining for comfort

They feature 100% cotton lining material.

Ordering information: Gloves are sold by the dozen pairs.

Caution: The degree of heat resistance a glove may offer is directly related to the duration of exposure and weight of the object being handled.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty