Would you like to log in again?
Terrycloth Baker's Pads, Wells Lamont
Supplier: Wells Lamont
B-PAD
80091-619EA 6.38 USD InStock
80091-619
Terrycloth Baker's Pads, Wells Lamont
Hot Hand Protectors
Heat-resistant terry cloth pads are designed for use in food service applications.
  • Two layers of heat-resistant terry cloth
  • Heavyweight material
  • Pads protect against up to 238 °C (460 °F)
  • For use in food service applications
The 24.1 cm long pad is constructed of two layers of terry cloth with a handhole.
All pads feature an elastic loop that allows them to be worn around the wrist or hung.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty