Would you like to log in again?
Terrycloth Sleeves, Wells Lamont®
Supplier: Wells Lamont
S-20MS S-15HR S-14HR S-11HR
80091-752EA 3.8 USD InStock
80091-752 80091-758 80091-756 80091-754
Terrycloth Sleeves, Wells Lamont®
Workwear
Cut and heat-resistant sleeves are available in medium weight or extra heavyweight terry cloth.
  • Cut and Heat-Resistant Sleeves
  • Medium and Extra Heavyweight Available
  • Terry Cloth
  • Elastic Tops
  • White
The medium-weight sleeve is seamless, features an elastic wrist, and resists temperatures up to 121°C (250°F)
All other sleeves have knit wrists. All sleeves are white and have elastic tops.
Extra-heavyweight sleeves resist temperatures up to 177°C (350°F). The 48.3 cm long sleeve features a hemmed wrist.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty