Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Standard Weight Y6243 Hot Mill Gloves, Wells Lamont

Supplier: Wells Lamont
Y6243L
80092-231PK 67.72 USD InStock
80092-231
Standard Weight Y6243 Hot Mill Gloves, Wells Lamont
Gloves
Standard weight Y6243 hot mill gloves protect up to 149°C (300°F).

  • Protects up to 300°F
  • 24 oz. natural cotton, nap-out
  • Straight thumb

These standard weight hot mill gloves protect hands from high heat, up to 300°F. Gloves are made of natural cotton with a straight thumb.

Ordering information: These gloves are sold by the dozen pair.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty