Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heavy Weight 30 oz. Hot Mill Gloves, Wells Lamont

Supplier: Wells Lamont
Y6302L
80092-241PK 80.91 USD InStock
80092-241
Heavy Weight 30 oz. Hot Mill Gloves, Wells Lamont
Gloves
This heavy-weight Hot Mill glove resists heat up to 400 °F.

  • Nap-out

The ambidextrous glove features a gauntlet safety cuff, straight thumb, and burlap lining.

Users can clean the glove in a washing machine.

Ordering information: Gloves are sold by the dozen pairs.

Caution: Caution Information: The degree of heat resistance a glove may offer is directly related to the duration of exposure and weight of the object being handled.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty