Would you like to log in again?
AlphaTec® 62-400 Fully Coated Natural Rubber Latex Gloves with Cotton Liner, Ansell
Supplier: Ansell
AlphaTec®
285653 285652 285654 285660
80093-112CS 957.98 USD InStock
80093-112 80093-110 80093-108 80093-106
AlphaTec® 62-400 Fully Coated Natural Rubber Latex Gloves with Cotton Liner, Ansell
Gloves
Flexible and durable chemical-protection gloves that are useful for a wide range of jobs.
  • Great flexibility and durability
  • Cotton liner keeps hands comfortable in hot or cold environments
  • Excellent grip for wet applications
  • Gloves stay flexible in cold temperatures
Fully coated natural rubber latex gloves offer great flexibility and durability.
Use in the chemical, fishing, food processing and service, general assembly, janitorial services, and petrochemical industries. Uses include chemical plants and transportation; chemical processing, oil refining and petrochemicals; fiberglass work; handling oils, acids, caustics and alcohols; plant maintenance; and seafood processing.
Soft-knit cotton liner helps keep hands and fingers comfortable in hot or cold environments. Gloves provide excellent grip for wet applications and excellent dexterity. Gloves stay flexible in cold temperatures (down to as low as -25°F, depending on contact time and conditions).
Certifications: Complies with FDA food handling requirements (21 CFR 177.2600). ANSI level 2 cut protection; ANSI Level 4 Abrasion protection.
Packaging: Gloves are packaged 12 pairs/polybag; 6 polybags/carton; 72 pairs/carton.
Caution: This product contains natural rubber latex, which may cause allergic reactions in some individuals.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty