Would you like to log in again?
AlphaTec™ 66-662 Chemical Protection Bib Overalls, Ansell
Supplier: Ansell
AlphaTec®
966410 966411 966409 966408 966412
80093-519EA 5677.22 USD InStock
80093-519 80093-517 80093-523 80093-521 80093-525
AlphaTec™ 66-662 Chemical Protection Bib Overalls, Ansell
Workwear
This lightweight, flexible, snag- and tear-resistant, stain-repellent and washable bib overall series provides outstanding chemical splash protection to a wide range of chemicals.
  • Chemical-resistant stitching and taped seams ensure superior performance
  • Tough and durable fabric
  • Anti-static properties
Breathable and lightweight Gore® chemical splash fabric transports moisture away from the body to reduce the risk of heat stress and provides all-day protection and comfort. Use in the chemical processing, general industry, and petrochemical industries. Uses include bulk chemical handling and transport, chemical plants and transportation, chemical processing, oil refining and petrochemicals, and handling specialty chemicals.
Certifications: Certified as Liquid Splash Protective Clothing for Hazardous material incidents in accordance with NFPA 1992.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty