Would you like to log in again?
B.O.D. Bottles, Bar-Coded, 300 ml, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
227497-02G 227497-10G 227497-04 227497-18G 227497-00G 227497-14G 227497-01 227497-03 227497-11G 227497-08 227497-17G 227497-09 227497-01G 227497-08G 227497-15G 227497-02 227497-12G 227497-14 227497-07G 227497-16 227497-17 227497-16G 227497-13 227497-06 227497-05 227497-06G 227497-00 227497-07 227497-18 227497-15 227498 227497-12 227497-04G 227497-03G 227497-10 227497-09G 227497-13G 227497-11 227497-05G
16285-209CS 558.94 USD InStock
16285-209 16285-207 16285-205 16285-203 16285-147 16285-125 16285-169 16285-201 74830-158 74830-156 74830-154 16285-140 74830-152 74830-174 74830-172 80093-756 16285-180 80093-758 74830-170 80093-752 80093-754 16285-114 16285-136 16285-158 74830-168 16285-199 74830-166 80093-760 16285-197 74830-164 16285-195 74830-162 16285-193 16285-170 80093-766 74830-160 16285-191 80093-762 80093-764
B.O.D. Bottles, Bar-Coded, 300 ml, WHEATON®, DWK Life Sciences
Bottles BOD Bottles
Incubation bottles are ideal for determining oxygen demand of sewage, water, and industrial wastes.
Borosilicate glass bottles are designed with large permanently screened-on labels for accuracy in recording bottle numbers. Interchangeable ground glass robotic stoppers and glass pennyhead stoppers are tapered to prevent air entrapment. Bottles fit most dissolved oxygen probes.
Diluted samples may be analyzed according to Standard Methods for the Examination of Water and Wastewater, 19th Ed., 5210B.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty