Would you like to log in again?
Rotors for Centrifuge 5430/5430 R
Supplier: Eppendorf
5427755004 022654161 022654047 022654403 5427754008 022654080 022654080 5427750002 022654209 022654403 022654004 022654128 5427750002 022654004 022654128 022654047 5427757007 022654306 022654209 022654161 022654306
80094-184EA 1191.05 USD InStock
CA80094-192 80094-184 89423-580 80094-192 CA80094-196 80094-188 80094-210 CA11028-850 80094-196 89239-160 CA80094-184 89423-578 CA80094-200 80094-200 CA80094-210 CA80094-188 CA80094-204 89423-576 80094-204 CA80094-208 80094-208
Rotors for Centrifuge 5430/5430 R
Centrifuges Rotors
Eppendorf rotors for centrifuge 5430/5430 R, precision at its core. Maximize your centrifuge's potential with rotors crafted for efficiency. Tailor your experiments with various capacities and speeds, ensuring optimal sample processing.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight and PTFE coated options available
Explore enhanced centrifugation possibilities with Eppendorf rotors designed for centrifuge 5430/5430 R. Choose from a range of capacities and speeds, providing versatility for microcentrifugation or larger sample volumes. The precision-engineered rotors ensure optimal sample separation. Accessories such as adaptors and buckets further customize your centrifuge setup. Maximize efficiency with the ability to handle diverse sample types. Elevate your laboratory's capabilities, achieve reproducible results, and enjoy the flexibility to meet specific application needs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty