Would you like to log in again?
VWR® Snap-On Label Holders for Wire Shelving
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
WR-1256 WR-1253
80094-272PK 61.47 USD InStock
80094-272 80094-274
VWR® Snap-On Label Holders for Wire Shelving
Label Holders
Flexible plastic holders snap into wire shelving with a 3.2 cm (11/4") face and accommodate inserts of the same width.
  • Flexible design
  • Durable plastic construction
  • Color: Clear
  • Replaceable inserts come in perforated sheets
Can be used to retrofit most major brands of new or existing wire shelving
Replacement inserts are laser and ink-jet printer compatible and provided in perforated 21.6Wx27.9Lcm (81/2x11") sheets.
Holders snap out for repositioning; inserts are easily exchanged. Color: clear.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty