Would you like to log in again?
VWR® Large Shelf Label Holders
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
APX-35 APX-46 APX-35M APX-46M
80094-286PK 118.5 USD InStock
80094-286 80094-284 80094-288 80094-290
VWR® Large Shelf Label Holders
Label Holders
Label holders are made of soft PVC that has been finished to eliminate glare for optimal bar-code readability.
  • Zero glare
  • Self-adhesive or magnetic holder options
  • Inserts are easily exchangeable
Label inserts are easily exchanged without removing the holder from its location
Replacement inserts are laser and ink-jet printer compatible.
Self-adhesive holders will adhere permanently to any surface. Magnetic holders are easily repositioned on metal surfaces.
Ordering information: Label holders are provided with inserts. Replacement inserts are supplied in 21.6Wx27.9Lcm (81/2x11") perforated sheets and include general formatting and printing instructions.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty