Would you like to log in again?
VWR® Shelf Label Holders
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
L-21 VWR L-11 VWR M-51 VWR L-51 VWR M-11 VWR L-31 VWR
80094-298PK 149.2 USD InStock
80094-298 80094-296 80094-302 80094-300 80094-306 80094-304
VWR® Shelf Label Holders
Label Holders
Label holders are made of semi-rigid clear butyrate plastic that has been finished to eliminate glare for optimal barcode readability.
  • Semi-rigid plastic
  • Easy to exchange
  • Magnetic or self-adhesive
  • Finish eliminates glare
Label inserts are easily exchanged without removing the holder from its location. Self-adhesive holders will adhere permanently to any surface. Magnetic holders are easily repositioned on metal surfaces.
Ordering information: Label holders are provided with inserts that are compatible with laser and ink jet printers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty