Would you like to log in again?
MicroSeal® SuperSorb® Sealed-Edge Cleanroom Wipers, Berkshire
Supplier: Berkshire
MicroSeal®
MSSS.0909.8 MSSS.0824.12 MSSS.1212.12
89095-084CS 1617.89 USD InStock
89095-084 94000-400 80094-525
MicroSeal® SuperSorb® Sealed-Edge Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Bonded 2-ply, 100% knitted polyester wipers feature superior absorbency with a high level of cleanliness and durability.
  • Critically low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, Acetone, and other solvents
  • Pinsonic design adds surface texture to aid in stubborn residue removal
They are ideal for applications where high concentrations of liquid are used and for cleaning and wiping rough surfaces. Recommended for ISO Class 3 (FED-STD-209E Class 1/M1.5) cleanroom environments, including semiconductor and biomedical cleanroom applications. Steam autoclavable for aseptic environments.
Ordering information: Wipers are laundered and packed in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Wipers come stacked in packs of 100.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty