Would you like to log in again?
Technologist Choice™ Zipper-Lock Bags, Bio Nuclear Diagnostics
Supplier: Bio Nuclear Diagnostics
Technologist Choice™
DIS-029 DIS-032 DIS-044 DIS-030 DIS-031 DIS-030P DIS-032 DIS-005 DIS-044
80094-728PK 120.08 USD InStock
80094-728 80094-736 CA40000-110 80094-730 80094-742 80094-740 80094-734 CA40000-108 80094-732
Technologist Choice™ Zipper-Lock Bags, Bio Nuclear Diagnostics
Bags Storage Bags
Technologist choice zipper-lock bags are made of 100% virgin raw material PE providing puncture resistance and clarity.
  • Single sheet with dual-side sealing for leak protection
  • Two-layer or four-layer bags available
  • Can be recycled
  • Resealable, avoid comtamination of inner packed items
  • 2-mil thickness offers regular strength
They are ideal for urine collection containers, blood tubes, or specimen transportation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty