Would you like to log in again?
Terminal Deoxynucleotidyl Transferase (TdT), EMD Millipore
Supplier: EMD Millipore
PF060-.155ML PF060-.155ML
80500-112EA 266.14 USD InStock
CA80500-112 80500-112
Terminal Deoxynucleotidyl Transferase (TdT), EMD Millipore
Enzymes
Biological Activity: Contains 775 units of TdT; one unit is the amount of enzyme required to transfer 1.0nmol of dAMP from dATP to the 3' -OH terminus of d(A)50 in 60 minutes at 37 °C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty