Would you like to log in again?
Calbiochem®, Phosphatase Inhibitor Cocktail Set II, EMD Millipore
Supplier: EMD Millipore
524625-1SET 524625-1SET
80501-130EA 558.87 USD InStock
80501-130 CA80501-130
Calbiochem®, Phosphatase Inhibitor Cocktail Set II, EMD Millipore
Cell Culture Supplements and Additives
A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs)
Suitable for use with cell lysates and tissue extracts, including samples containing detergents.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty