Would you like to log in again?
Proteinase K
Supplier: EMD Millipore
71049-4 71049-4 71049-4 71049-3
102969-030EA 444.86 USD InStock
102969-030 CA80509-758 80509-756 80509-758
Proteinase K
Enzymes
Proteinase K is a highly active 28,904-Da serine protease isolated from the fungus Tritirachium album.
The enzyme exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA. Its activity is increased in the presence of denaturants such as SDS (1%) and elevated temperature (50 to 60 °C). Working concentration is 50 to 100 µg/ml for protein removal and enzyme inactivation and upto 2 mg/ml for tissue treatment. It is a lyophilized powder.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty