Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Rosetta™ Competent Cells, Expression Strains, MilliporeSigma
Supplier: MilliporeSigma
70956-4 70956-3 70956-4 70953-4 70954-3 70953-4 70954-4 70956-3 70953-3 70953-3 70954-4 70954-3
80509-776EA 237.88 USD InStock
80509-776 CA80509-790 80509-770 CA80509-770 CA80509-768 80509-790 80509-788 80509-778 CA80509-776 CA80509-788 80509-768 CA80509-778
Rosetta™ Competent Cells, Expression Strains, MilliporeSigma
Competent Cells
Rosetta™ host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
  • tRNA genes are driven by their native promoters
  • Standard for 20 or 50 reactions
  • Guaranteed efficiency: >2×106 cfu/µg for host strains
These strains supply tRNAs for AGG, AGA, AUA, CUA, CCC, GGA codons on a compatible chloramphenicol-resistant plasmid. Thus the Rosetta™ strains provide for 'universal' translation which is otherwise limited by the codon usage of E. coli.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lac UV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. In Rosetta(DE3)pLysS, the rare tRNA genes are present on the same plasmids that carries the T7 lysozyme gene.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty