Would you like to log in again?
KOD DNA Polymerase, MilliporeSigma
Supplier: MilliporeSigma
71085-3 71085-3
80511-382EA 701.52 USD InStock
80511-382 CA80511-382
KOD DNA Polymerase, MilliporeSigma
Nucleic Acid Reagents End-point PCR Enzymes and Kits
KOD DNA Polymerase is a recombinant form of Thermococcus kodakaraensis KOD1 DNA polymerase.
  • Higher fidelity, excellent for cloning
  • Greater yield, its extension speed is fast
  • Amplifies plasmid and lambda DNA upto 6 kbp and genomic DNA upto 2 kbp
  • No truncated amplification products
It is a high fidelity thermostable DNA polymerase that amplifies target DNA up to 6 kbp with superior accuracy and yield for PCR applications. The enzyme's 3'→5' exonuclease-dependent proofreading activity results in a lower PCR mutation frequency than any other available DNA polymerase.
The elongation rate and processivity are 5 times and 10 to 15 times higher, respectively, than for Pfu DNA polymerase, resulting in highly accurate and robust yield, in a short reaction time. The enzyme generates blunt-ended PCR products suitable for cloning with various vector kits.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty