Would you like to log in again?
NovaTaq™ Hot Start DNA Polymerase, MilliporeSigma
Supplier: MilliporeSigma
NovaTaq™
71091-3 71091-4 71091-4 71091-3
80511-392EA 688.09 USD InStock
CA80511-394 80511-392 CA80511-392 80511-394
NovaTaq™ Hot Start DNA Polymerase, MilliporeSigma
Nucleic Acid Reagents End-point PCR Enzymes and Kits
NovaTaq™ Hot Start DNA Polymerase is a chemically modified form of Taq DNA polymerase that is inactive at ambient temperature.
  • Highly specific, thereby minimizes the non-specific amplifications
  • Improved low-copy target amplification
  • Ambient temperature setup compatible with automation
  • Target amplification of up to 5 kbp
  • Ideal for quantitative and high-throughput PCR applications
The enzyme provides improved specificity when compared to standard Taq DNA polymerase and can minimize the generation of nonspecific amplification products, such as primer-dimers and misprimed products.
The enzyme must be activated by heat treatment (7 - 10 min at 95 °C), after which thermal cycling can proceed. The enzyme generates PCR products with 3′-nucleotide overhangs, suitable for cloning with a varierty of vector kits.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty