Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Rosetta-gami™ B Competent Cells, Expression Strains, EMD Millipore
Supplier: EMD Millipore
71136-3 71135-3 71137-4 71135-3 71136-4 71137-3 71137-4 71135-4 71135-4 71136-3 71137-3 71136-4
80601-670EA 173.93 USD InStock
80601-670 CA80601-680 80601-680 CA80601-670 80601-672 80601-674 CA80601-674 80601-676 CA80601-672 80601-678 CA80601-678 CA80601-676
Rosetta-gami™ B Competent Cells, Expression Strains, EMD Millipore
Competent Cells
Rosetta-gami B strains combines the key features of BL21 (and its Tuner™ derivative), Origami, and Rosetta to enhance both the expression of eukaryotic proteins and the formation of target protein disulfide bonds in the bacterial cytoplasm.
  • T7 lysozyme expression suppresses basal T7 expression
  • Compatible with ampicillin- or spectinomycin-resistant vectors
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty