Would you like to log in again?
Protease (from Streptomyces griseus)
Supplier: EMD Millipore
537088-100KU 537088-5KU 537088-5KU 537088-25KU 537088-50KU 537088-25KU 537088-50KU 537088-10KU 537088-10KU
80602-616EA 562.15 USD InStock
CA80602-616 80602-616 CA80602-618 80602-614 80602-612 80602-610 CA80602-612 CA80602-614 80602-618
Protease (from Streptomyces griseus)
Enzymes
A non-specific protease liquefies mucins and digests proteins to free amino acids.
  • Can be used to isolate living chondrocytes
  • Most active at neutral pH, but stable over wide range of pH and temperature
  • Lyophilized form
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty