Would you like to log in again?
Jar Rolling Mills, Bench, Sepor
Supplier: Sepor
010E-035 110V/60HZ 010E-027
80700-006EA 35543.05 USD InStock
80700-006 80700-004
Jar Rolling Mills, Bench, Sepor
Mills
These continuous duty jar drives are used for batch wet or dry grinding, blending, dispersing or other conditioning.
  • Roll spacing is adjustable to accommodate jar sizes of 2 to 15" dia.
  • All models have free wheeling jar stops
  • Constructed of highest quality materials
Variable speed DC drive for an output range of 20 to 345 roller RPM; thus an 8 inch diameter jar would have a speed range of 5 to 86 RPM. Water tight electrical system and totally enclosed gear motor. These mill drives are built to last with very little maintenance and can drive heavy, fully charged mills with ease.
Ordering information: Specify 115V/1Ph/60 Hz or 230V/1Ph/50Hz. Jars sold separately. Optional accessories include revolution counters and automatic timers; contact your Avantor representative for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty