Would you like to log in again?
Erlenmeyer Flask, Polypropylene, Dynalon
Supplier: Dynalon
241365 241345
30621-006CS 179.01 USD InStock
30621-006 80700-012
Erlenmeyer Flask, Polypropylene, Dynalon
Flasks Erlenmeyer Flasks
Autoclavable polypropylene Erlenmeyer flask with molded graduations offers excellent chemical resistance.
Graduations are permanently molded into the flask.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty