Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium fluoride

Supplier: Avantor

ANAC029-941-01
80700-148EA 181.72 USD InStock
80700-148
Calcium fluoride
Calcium fluoride


Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500 °C
Density: 3.18 g/ml
MDL Number: MFCD00010907
CAS Number: 7789-75-5
Order Now
Specification Test Results
Quality level 100
Grade Certified reference material
Format Matric material

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty