Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium nitrate, crystals

Supplier: Avantor

ANACFA209A-22.7KG
80700-150EA 338.8 USD InStock
80700-150
Potassium nitrate, crystals
Potassium nitrate


Formula: KNO₃
MW: 101.1 g/mol
Melting Pt: 334 °C
MDL Number: MFCD00011409
CAS Number: 7757-79-1
Order Now
Specification Test Results
Grade ACS reagent
Quality level 200
Assay ≥99.0%
Impurities ≤0.005% insolubles
pH 4.5 - 8.5

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty