Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium nitrate, powder

Supplier: Avantor

ANAC030-025-01
80700-170EA 301.84 USD InStock
80700-170
Sodium nitrate, powder
Sodium nitrate


Formula: NaNO₃
MW: 84.99 g/mol
Melting Pt: 306 °C
MDL Number: MFCD00011119
CAS Number: 7631-99-4
Order Now
Specification Test Results
Assay ≥99.0%
pH 5.5 - 8

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty