Would you like to log in again?
CRUCIBLE ZIRCONIUM LF 25ML
Supplier: METAL TECHNOLOGY INC.
Zirconium Crucibles 25ml^ Zirconium Crucibles, Low form, Corrosion resistance in most organic and inorganic acids, Salt solutions, Strong alkalis and a few molten salts, Passive oxide film, Good in both acid and base solutions, Depth: 23mm, Bottom diamete
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRUCIBLE ZIRCONIUM LF 25ML 80700-502 10-0025LF Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty