Would you like to log in again?
CONICAL BUCKET 8X50ML SET/4
Supplier: Thermo Fisher Scientific
Conical Bucket 8x50ml^ Conical Bucket, Unsealed, No adapter needed, Maximum tube dimension: 29.5 x 120mm, Maximum speed: 4,500 rpm, Maximum RCF: 3,260xg, Size: 8x50 mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONICAL BUCKET 8X50ML SET/4 80700-978 75005703 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty