Would you like to log in again?
Ball Peen Hammers, Anachemia
Supplier: Anachemia
45RC15 45RC16
80701-366EA 35.03 USD InStock
80701-366 80701-368
Ball Peen Hammers, Anachemia
Mallets and Hammers
Ball peen hammers are available in bead and button styles, for a variety of tasks.
The ball peen bead hammer is ideal for flattening beads. It features a durable steel shaft and a rubber grip handle for maximum comfort.
The ball peen button hammer is ideal for deslagging. It features a steel shaft and rubber grip handle for dependable, long-lasting use and maximum comfort.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty