Would you like to log in again?
Heat Treating Furnace, Model C163210/PM3T, Cress
Supplier: Cress
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FC163210/PM3-23
80701-408EA 25610.28 USD InStock
80701-408
Heat Treating Furnace, Model C163210/PM3T, Cress
Ovens Furnaces (Ashing and Muffle Ovens)
The model C163210/PM3T is a heat treating furnace used for treating metals with a maximum temperature of up to 2250°F.
  • Features a Watlow PM3T single set point with timer
The chamber is constructed of firebrick of the best quality. It measures 2.5” and it is backed with 1.5” of 1800°F block insulation. The furnace shell is 18 GA. sheet metal. The door opens up away from the user. There is a rope seal on the door that provides a better seal against the furnace. The door mechanism uses two heavy duty springs that keep the door safely closed or open.
Ordering information: This unit is available in 240V, 208V. 480V is an additional charge. Optional PM4 controller with double display is available; it has four files that can hold up to 10 steps each file. Contact your Avantor representative for more information.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty