Would you like to log in again?
MetroMax i™ Mobile Desk, Metro™
Supplier: Metro International
MetroMax™
MDSX3
82000-938EA 2662.25 USD InStock
82000-938
MetroMax i™ Mobile Desk, Metro™
Carts
Mobile desk can be used for storage or can be combined with accessories to provide a multi-functional lab desk
Customize your desk with slide-out keyboard, flat monitor mounting bracket, and power strip. Keyboard shelf is epoxy-coated for a high level of corrosion resistance. Bracket mounts flat computer monitor between two posts on desk. Six-outlet power strip features surge protection and mounts flush with shelf frame. Power strip is UL listed.
Phenolic work surface offers a high level of chemical- and corrosion-resistance. The longitudinal beams and posts are made from continuous glass fibers and thermoset resin composite material. Ledges, dividers, and wedge are made from glass-reinforced polypropylene. The post and center beam caps are made from high-density polyethylene.
Ordering information: Four locking 12.7cm (5") polyurethane brake casters included for maximum stability. Manufacturer’s lifetime guarantee against corrosion and rust. Accessories sold separately. Power strip includes 1.8m (6') cord.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty