Would you like to log in again?
Super Erecta Shelf® Lab Security Cart, Metro™
Supplier: Metro International
Super Erecta Shelf®
SECMLAB SECMLAB
82000-940EA 3037.9 USD InStock
CA82000-940 82000-940
Super Erecta Shelf® Lab Security Cart, Metro™
Carts
Stainless steel security cart with flip-up door provides secure storage for animal cage, lab instrument, or hazardous chemical transport
Features two stainless steel wire shelves, one stainless steel security module, and one stainless steel solid shelf that can be used as a work surface and features a raised ship's edge to contain unwanted spills. Super Erecta Shelf® construction offers ease of assembly, absolute rigidity, and quick and simple relocation of wire shelves. Open construction minimizes dust accumulation and maximizes visibility and air circulation.
Ordering information: Cart includes four donut bumpers and four 12.7cm (5") polyurethane casters (2 swivel, 2 brake).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty