Would you like to log in again?
MetroMax™ General Task Station, Metro™
Supplier: Metro International
MetroMax™
CSX3
82000-952EA 992.53 USD InStock
82000-952
MetroMax™ General Task Station, Metro™
Carts
General task station can be used for benchtop instrument or equipment storage, or can be combined with accessories to create a computer task station
Slide-out keyboard shelf, flat monitor mounting kit, and power strip can be added to create a computer task station. Keyboard shelf is epoxy-coated for high level of corrosion resistance. Flat monitor mounting kit consists of a mounting bracket and a cross bar, which permit a flat monitor to be mounted between the task station posts. Six-outlet power strip with surge protection mounts flush with the shelf frame and includes a 1.8m (6') cord.
Designed to maximize your benchtop space. Station is comprised of 83.8cm (33") posts, 2 solid shelves, 2 side ledges, 1 back ledge, and 4 nonskid leveling feet. Features strong polymer construction with superior corrosion resistance. Solid shelves contain spills and are adjustable in 1.3cm (1/2") increments along the height of the posts. Stainless steel leveling bolts and nylon covers make leveling feet an effective nonskid solution.
Certifications: Power strip is UL listed.
Ordering information: Manufacturer's lifetime warranty against corrosion and rust.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty