Would you like to log in again?
Monoject™ SoftPack Syringes, Sterile, Covidien
Supplier: Covidien
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Monoject™
1183500888 1180600777 Z1182000777 1180300777 1180325114 1183500555 1180300555 Z1180600555 BD3095701 1182000777 1180322100 1182000555 1181621112 1180600555 1180325058 Z8881520673 1181621100 1183500777 1181621100 1180321100 1180322112 Z1183500777 1180327114 1180320100 1181620112 1181622112 1180321112 1180300777 1180323100 1181620112 1180320112
82002-244CS 191.48 USD InStock
82002-244 CA82002-299 CA82002-310 82002-301 82002-268 82002-246 82002-264 82002-260 CA82002-304 CA82002-306 CA82002-246 CA82002-268 82002-258 82002-254 82002-299 82002-310 82002-278 82002-256 82002-312 82002-272 82002-250 14222-240 82002-274 82002-296 82002-252 82002-297 82002-306 82002-308 82002-302 CA82002-278 82002-304
Monoject™ SoftPack Syringes, Sterile, Covidien
Syringes Syringes, Medical
These latex-free syringes feature a one-step peel-away package that facilitates quick and easy opening for aseptic handling.
The 3mL syringes feature extended finger flanges and a longer plunger rod for greater control. The 3mL, 6mL, and 12mL syringes feature bold graduations; the 35mL and 60mL syringes feature bold dual graduations. The 35mL syringes also feature 1oz. graduations in 0.25oz. increments, and the 60mL syringes have 2oz. graduations in 0.25oz. increments.
Each individual package contains a color-coded gauge stripe and bar code for easy gauge identification. Syringes feature clear barrel construction for precise medication measurement and easier visualization of flashback. Syringes are available with or without sterile, ultra-sharp, tri-beveled, anti-coring needles. Needles feature improved lubricant for increased patient comfort. Syringes with needles feature a Luer-lock tip. Syringes are autoclavable in the rigid pack for use in O.R./procedure packs.
Ordering information: Tip caps are sold separately; for more information contact your Avantor representative.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty