Would you like to log in again?
SmartTray™ System, Metro™
Supplier: Metro International
SmartTray™
CBNTC30M CBNTC30MSOL2 CBNTC20M CBNTCS20M CBNTCS20M CBNTC20MSOL2 CBNTC30M CBNTC20MSOL1 CBNTC20MSOL1 CBNTC20M CBNTC30MSOL2 CBNTC30MSOL1 CBNTC20MSOL2 CBNTC30MSOL1
82002-430EA 7180.95 USD InStock
82002-430 CA82002-450 CA82002-430 82002-450 CA82002-448 82002-428 CA82002-436 CA82002-428 82002-434 82002-446 82002-436 CA82002-446 82002-448 CA82002-434
SmartTray™ System, Metro™
Carts
Designed to maximize productivity by accelerating throughput and minimizing PCB handling damage
Premium ESD tray inlay (22221-396) offers maximum ESD protection in addition to superior resistance to heat, abrasion, and chemicals. The soft dissipative top layer makes it ideal for use in soldering and assembly areas. Economy tray inlays are not recommended for use in soldering applications.
This cart and tray system facilitates efficient tracking of work-in-process and finished goods with the use of bar code technology. Carts and trays can also be color-coded for easy sorting and categorizing through the use of colored labels (not included). A detent system allows pulled-out trays to be suspended from the cart for optimum ergonomic access. Molded from Metrostat™ material, system provides premium ESD protection for sensitive electronic parts. SmartTray trays can be used with all bottom-load tray cart designs, Metro and non-Metro models, as well as conveyor systems.
Ordering information: Each cart comes complete with four vibration suppression casters (two brake and two swivel), a grounding cable, and two stop bars. Short SmartTray carts include two push handles. Color-coding labels are available; contact your Avantor representative for more information.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty