Would you like to log in again?
Super Erecta® Wire Shelving, Super Erecta Brite® Finish, Metro™
Supplier: Metro International
Super Erecta™ Super Erecta®
1836BR 1836BR 1848BR 2448BR 2448BR 2436BR 1848BR 2436BR
82002-474EA 107.37 USD InStock
82002-474 82002-476 CA82002-474 CA82002-480 82002-480 82002-478 CA82002-476 CA82002-478
Super Erecta® Wire Shelving, Super Erecta Brite® Finish, Metro™
Furniture Shelves
Design your own wire shelf system with appropriate accessories.
  • The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap
Note: The actual length of the shelves is 3.2 mm (¹/₈") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6cm high are not for use with 35.6 cm wide shelves.
Super Erecta Brite® shelves are made of zinc-plated steel. Open-wire design minimizes dust accumulation, allows a free circulation of air, and provides greater visibility of stored items. Easy shelf adjustment is possible through the use of SiteSelect® chrome-plated stationary or mobile posts, which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1") increments along the entire height of the post. An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Certifications: Shelves meet MIL-S-40144E specifications.
Ordering information: Each shelf includes four pairs of plastic split sleeves for securing shelf to posts.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty