Would you like to log in again?
Tak Pak® 712™ Accelerator, Loctite®, Henkel
Supplier: Henkel
Tak Pak®
135316 229783 229783 135265
82002-781CS 1226.8 USD InStock
82002-781 82002-758 82002-764 82002-766
Tak Pak® 712™ Accelerator, Loctite®, Henkel
Adhesives
LOCTITE® SF 712™ Accelerator increases cure speed of Loctite instant adhesive
  • Isopropanol base
  • Excellent for cosmetic applications, for clear and transparent adhesive beads or fillets
  • Suitable for securing wires or coils to PCBs
Typical applications include securing wires or coils to PCBs, tamper-proofing adjustable components, mounting stand-offs, edge guides and board stiffeners.
It is suited for post-application on cyanoacrylate
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty