Would you like to log in again?
UV CURING ADHSV 4000CP 25ML
Supplier: LOCTITE CORP
LiteTak* 3751 Light Cure Adhesive^ Clear/colorless UV cure product. Primary use is tacking and sealing on a variety of substrates. Viscosity of 4,000 cP at 25[degree]C. This product is designed for applications where rapid cure and a dry to the touch surface are desired. This is solvent free material.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
UV CURING ADHSV 4000CP 25ML 82002-786 135321 Case of 10 Retrieving Restricted
Restricted To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty