Would you like to log in again?
VWR® Extruded Natural Rubber Tubing
Supplier: VWR
10351
82003-352EA 724.35 USD InStock
82003-352
VWR® Extruded Natural Rubber Tubing
Tubing Transfer Tubing
The typical physical properties of this natural rubber tubing closely match natural rubber latex.
  • Excellent tear resistance
  • High abrasion resistance
  • Wide range of temperature tolerances
  • Available in a wide variety of standard sizes
Natural rubber tubing is produced from dry natural rubber.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty