Would you like to log in again?
Conductive Divider Boxes, LEWISBins+™, Orbis
Supplier: Orbis
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DC2050XL DC2035XL DC2070XL
82003-494EA 28.76 USD InStock
82003-494 82003-492 82003-498
Conductive Divider Boxes, LEWISBins+™, Orbis
Boxes ESD-Safe Boxes
Permanently conductive containers are made of a strong and durable carbon-filled, injection-molded polypropylene copolymer resin
Ideal for use in commercial, industrial, electronics, and research facilities for storage, assembly, fabrication, and distribution applications. Compatible with many types of racking, shelving, and material handling equipment. Boxes feature a strong stacking ridge and ribbed walls. Two cardholder snaps are standard on each box. Color: black.
Ordering information: Dividers and covers are also available, contact your VWR representative for more information.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty