Would you like to log in again?
Plastibox® Conductive Bins, LEWISBins+™
Supplier: Orbis
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Plastibox®
PB31XL PB40XL PB30XL PB22XL PB50XL PB20XL
82003-518CS 288.65 USD InStock
82003-518 82003-520 82003-512 82003-522 82003-516 82003-514
Plastibox® Conductive Bins, LEWISBins+™
Boxes ESD-Safe Boxes
ESD-Safe Part Bins are permanently molded in conductive material for use in cleanrooms and workstations.
  • Different sizes available
  • Protect parts from the damaging effects of static electricity
  • Label insert area for easy identification
  • Smooth gravity flow interior optimizes parts accessibility
  • Bins hang on most louvered panels or rails
Open, sloping hopper fronts for easy product removal. These stackable bins also feature rear hanging lips for attachment to most louvered panels or rails.
Combine Part Bins with ESD-Safe Metal Storage Systems to meet your work-in-process requirements.
Color: Black
Ordering information: Minimum order of 24.
Caution: Note: PB50 is not designed for use on hanging systems.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty