Would you like to log in again?
COVERALL EXTRA WHT MED CS25
Supplier: KIMBERLY CLARK CORP
KLEENGUARD* Extra Coveralls^ Extra durability for applications requiring fine particle protection. Microforce* fabric layer gives premium SMS fabric added particle holdout, yet allows excellent breathability. Zipper front with 2.5cm flap. Elastic back, wrists, ankles. Color: white. Size: medium.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVERALL EXTRA WHT MED CS25 82003-876 46102 Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty