Heavier than KLEENGUARD® A20, these coveralls provide extra durability for applications requiring fine particle protection.
- Provide durable chemical splash and fine particle protection
- 25 x more breathable than Tyvek® yet maintains comparable particle holdout
- Cloth-like and abrasion-resistant outer layer
- Microfiber middle layer filters out many water-based liquids and dry particulates
- Feature zipper front with flap and elastic back
Breathable MICROFORCE™ Barrier SMS fabric has outer layers of abrasion-resistant spunbond polypropylene and middle layers of an intricate web of microfibers that filter out many water-based liquids and dry particulates
Fabric meets Class 1 Normal Flammability under Federal Flammable Fabrics Act (CPSC 1610). Fabric will melt when exposed to open flame or temperatures over 150 °C (302 °F). Coveralls are NFPA 99 compliant for antistatic materials.
Fabric is 25 x more breathable than Tyvek® material, yet it maintains comparable particle holdout. Zipper front with 2.5 cm (1") flap