Would you like to log in again?
Prinzing® Ball Valve Lockouts, Brady Worldwide®
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Prinzing®
BS08A-YW BS08A BS07A-RD BS08A-YW 45348 BS08A-BU BS07A-BU 45344 BS07A-BU BS08A-BU BS08A-GN BS08A-GN BS07A-YW
82004-220EA 69.81 USD InStock
82004-220 CA82004-220 82004-222 CA82004-218 CA82004-222 82004-224 CA82004-224 82004-226 CA82004-226 82004-228 CA82004-228 82004-218 CA82004-216
Prinzing® Ball Valve Lockouts, Brady Worldwide®
Security Locks
Designed to lock a wide range of valves, these small and large ball valve lockouts are both versatile and easy to use
Two halves encompass the ball valve handle to secure it from inadvertent use. Lockouts are made from ultra-tough polypropylene.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty