Would you like to log in again?
CONFORM® Stretch Bandages, Covidien
Supplier: Covidien
CONFORM®
2231 2242 2232 2236
82004-748CS 298.01 USD InStock
82004-748 82004-750 82004-744 82004-746
CONFORM® Stretch Bandages, Covidien
First Aid Supplies Wound Dressings
One-ply cotton and polyester-blend crocheted bandages secure dressings, I.V.s, and splints.
  • Length: 3.7 m (4⅟₁₀ yd.)
  • Low linting
  • Absorbent
  • Minimal taping needed
  • Cotton and polyester blend
They can also provide mild compression or support.
Bandages hold securely to any body contour for movement.
Packaging: Nonsterile bandages are bulk packaged. Sterile bandages are individually packaged in soft pouches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty